missing translation for 'onlineSavingsMsg'
Learn More

Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™

Product Code. 10467963
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail

Glycine (White Crystals or Crystalline Powder)

Glycine is used in the preparation of Tris–Glycine electrophoresis buffers and culture media.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 182°C
Absorbance 0.1 AU max. at 280nm
Ammonium 0.02% max.
Arsenic (As) 1ppm max.
pH 5.9 to 6.4
Color White
Chloride 0.003% max.
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 98.5 to 101.0 %
Ignition Residue 0.10% max.
Identification Pass Test
Iron (Fe) 7ppm max.
Loss on Drying 0.20% max.
Merck Index 15, 4526
Other Amino Acids Pass Test
Packaging Poly Bottle
Formula Weight 75.07
Percent Purity 98.5 to 101.0%
Density 1.595g/cm³
Quantity 500 g
Physical Form Powder
Chemical Name or Material Glycine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'