missing translation for 'onlineSavingsMsg'
Learn More

Glycolate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™

Product Code. 12952125
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

Molecular Formula C2H3O3-

Specifications

Concentration 1g/L
Packaging Bottle
Quantity 250 mL
Type Single-component Standard Solution
Solvent or Matrix H2O
Physical Form Liquid
Chemical Name or Material Glycolate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'