missing translation for 'onlineSavingsMsg'
Learn More
Glycoluril, 97%, Thermo Scientific Chemicals Product Code.: 15565036

Glycoluril, 97%, Thermo Scientific Chemicals

Product Code. 15565036
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15565036

Brand: Thermo Scientific Acros 120130250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-46-8
Molecular Formula C4H6N4O2
Molecular Weight (g/mol) 142.12
InChI Key VPVSTMAPERLKKM-UHFFFAOYSA-N
Synonym glycoluril, acetyleneurea, acetylenediureine, acetylenediurea, glyoxalbiuret, glyoxaldiureine, acetylene carbamide, diurea glyoxalate, tetrahydroimidazo 4,5-d imidazole-2,5 1h,3h-dione, glyoxaldiureneShow MoreShow Less
PubChem CID 62347
ChEBI CHEBI:42946
IUPAC Name 1,3,3a,4,6,6a-hexahydroimidazo[4,5-d]imidazole-2,5-dione
SMILES C12C(NC(=O)N1)NC(=O)N2

Specifications

Melting Point >300.0°C
Boiling Point 862.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (N)
Quantity 25 g
Beilstein 26,441
Merck Index 15,88
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in warm mineral acids and in warm ammonia
Formula Weight 142.12
Percent Purity 97%
Chemical Name or Material Glycoluril
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465672 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number120131000,120135000,120130250
Lot NumberA0455670 Certificate TypeCertificate of Analysis Date28/08/2023 Catalog Number120131000,120135000,120130250
Lot NumberA0439808 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number120131000,120135000,120130250
Lot NumberA0397061 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number120131000,120135000,120130250
Lot NumberA0404542 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number120131000,120135000,120130250
compliance-icons
Product Identifier
  • Glycoluril
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycoluril, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.