missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Glycylglycine, 99+%
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
Specifications
Specifications
Melting Point | 262.0°C to 264.0°C |
Absorbance | (10% aq. soln.) (1 cm cell), 0.01 max. at 405nm |
pH | 5.5 to 6 (5% aq. soln.) |
Color | White |
Chloride | 100ppm max |
Assay Percent Range | 99+% |
Infrared Spectrum | Authentic |
Linear Formula | H2NCH2CONHCH2CO2H |
Beilstein | 04, 371 |
Loss on Drying | 0.5% max. (105°C, 4 hrs) |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'