Learn More
Thermo Scientific Chemicals Glyoxal, pure, 40 wt.% solution in water
39.09 € - 707.52 €
Specifications
Chemical Name or Material | Glyoxal |
---|---|
Name Note | 40 wt.% Solution in Water |
CAS | 107-22-2, 7732-18-5 |
Melting Point | -14.0°C |
Color | Colorless to Yellow |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10582551
|
Thermo Scientific Chemicals
156225000 |
500 mL | Glass bottle |
39.09 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10081700
|
Thermo Scientific Chemicals
156220010 |
1 L | Glass bottle |
64.92 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10091700
|
Thermo Scientific Chemicals
156220025 |
2.5 L | Glass bottle |
126.63 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10593314
|
Thermo Scientific Chemicals
156220250 |
25 L | Metal Drum |
707.52 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Glyoxal | |
107-22-2, 7732-18-5 | |
Colorless to Yellow | |
104.0°C | |
C2H2O2 | |
MFCD00006957 | |
01,413 | |
1.265 | |
Solubility in water: miscible. | |
C(=O)C=O | |
58.04 | |
CHEBI:34779 | |
58.04 | |
Pure |
40 wt.% Solution in Water | |
-14.0°C | |
1.2650g/mL | |
>104°C | |
HCOCHO | |
01, 759 | |
15, 4544 | |
glyoxal, ethanedial, oxalaldehyde, 1,2-ethanedione, glyoxylaldehyde, biformal, biformyl, diformal, diformyl, aerotex glyoxal 40 | |
LEQAOMBKQFMDFZ-UHFFFAOYSA-N | |
oxaldehyde | |
7860 | |
8 mPa.s (20°C) | |
39 to 41% (Titrimetry other) | |
Liquid |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects if inhaled.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritatio
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-474-9
Recommended Storage : May darken during storage
- Glyoxal, 40 wt% solution in water
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H332-Harmful if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.