Learn More
Glyoxylic Acid 50% in Water, Thermo Scientific Chemicals
21.40 € - 173.00 €
Chemical Identifiers
CAS | 298-12-4, 7732-18-5 |
---|---|
Molecular Formula | C2H2O3 |
Molecular Weight (g/mol) | 74.035 |
MDL Number | MFCD00006958 |
InChI Key | HHLFWLYXYJOTON-UHFFFAOYSA-N |
Synonym | glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid |
PubChem CID | 760 |
ChEBI | CHEBI:16891 |
IUPAC Name | oxaldehydic acid |
SMILES | C(=O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10095720
|
Thermo Scientific Acros
411060050 |
5 g | Glass bottle |
21.40 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10187840
|
Thermo Scientific Acros
411065000 |
500 g | Glass bottle |
57.10 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10300194
|
Thermo Scientific Acros
411060025 |
2.5 kg | Glass bottle |
173.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
298-12-4, 7732-18-5 | |
74.035 | |
HHLFWLYXYJOTON-UHFFFAOYSA-N | |
760 | |
oxaldehydic acid |
C2H2O3 | |
MFCD00006958 | |
glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid | |
CHEBI:16891 | |
C(=O)C(=O)O |
Specifications
Colorless to Yellow | |
111.0°C | |
C2H2O3 | |
MFCD00006958 | |
05,320; 07,162; 09,228 | |
15, 4546 | |
Solubility in water: miscible. | |
C(=O)C(=O)O | |
74.035 | |
CHEBI:16891 | |
≥50% | |
1.3 | |
Glyoxylic acid, 50% in water |
298-12-4, 7732-18-5 | |
-93.0°C | |
HC(O)COOH | |
03, IV, 1489 | |
1.3000g/mL | |
glyoxylic acid, 2-oxoacetic acid, oxoacetic acid, oxoethanoic acid, glyoxalic acid, formylformic acid, acetic acid, oxo, oxalaldehydic acid, glyoxylate, alpha-ketoacetic acid | |
HHLFWLYXYJOTON-UHFFFAOYSA-N | |
oxaldehydic acid | |
760 | |
74.04 | |
1.4140 to 1.4180 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause an allergic skin reaction.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 206-058-5
missing translation for 'rtecsNumber' : MD4550000
missing translation for 'tsca' : TSCA
RUO – Research Use Only