missing translation for 'onlineSavingsMsg'
Learn More

Glyoxylic acid monohydrate, 98%, pure, Thermo Scientific Chemicals

Product Code. 10647652
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-96-2
Molecular Formula C2H2O3·H2O
Molecular Weight (g/mol) 92.06
MDL Number MFCD00127974
InChI Key MOOYVEVEDVVKGD-UHFFFAOYSA-N
Synonym glyoxylic acid monohydrate, glyoxylic acid hydrate, 2-oxoacetic acid hydrate, oxaldehydic acid hydrate, glyoxalic acid monohydrate, glyoxalate, glyoxylate hydrate, glyoxalic acid hydrate, oxoacetate monohydrate, glyoxylic acid-hydrate, oxoacetic acid hydrate
PubChem CID 15620607
IUPAC Name oxaldehydic acid;hydrate
SMILES C(=O)C(=O)O.O

Specifications

Melting Point 48.0°C to 52.0°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula HCOCO2H·H2O
Quantity 100 g
Solubility Information Solubility in water: freely soluble.
Formula Weight 92.06
Percent Purity 98%
Grade Pure
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material Glyoxylic acid monohydrate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Glyoxylic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.