Learn More
Sampling Systems™ GMP Pallet Knife
These stainless steel GMP pallet knives are ideal for a wide range of applications, including cleaning, scraping and mixing. Made from medical grade 420 stainless steel, they are crevice-free and have a polished finish for easy cleaning after use.
Brand: Sampling Systems™ A123-250
This item is not returnable.
View return policy
Description
- Strong and flexible
- Range of different lengths
Specifications
Non-sterile | |
Curved | |
Pallet Knife | |
380 mm |
Autoclavable | |
Reusable | |
Stainless Steel |