missing translation for 'onlineSavingsMsg'
Learn More

GMPS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Bio-Techne NBP1-52972

 View more versions of this product

Product Code. 18292375

  • 453.55 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GMPS Polyclonal specifically detects GMPS in Human samples. It is validated for Western Blot.
Specifications

Specifications

GMPS
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
EC 6.3.5.2, Glutamine amidotransferase, GMP synthase, GMP synthase [glutamine-hydrolyzing], GMP synthetase, guanine monphosphate synthetase, guanosine 5'-monophosphate synthase, MLL/GMPS fusion protein
Rabbit
77 kDa
100 μL
metabolism
8833
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
P49915
GMPS
Synthetic peptides corresponding to GMPS(guanine monphosphate synthetase) The peptide sequence was selected from the middle region of GMPS. Peptide sequence VCTALLNRALNQEQVIAVHIDNGFMRKRESQSVEEALKKLGIQVKVINAA.
Affinity purified
RUO
Primary
Expected identity based on immunogen sequence: Chicken: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Bovine: 92%; Guinea pig: 92%; Zebrafish: 92%.
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers