missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ GMPS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NBP3-29817

Product Code. 30045512

  • 556.14 € / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GMPS Polyclonal antibody specifically detects GMPS in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications

Specifications

GMPS
Polyclonal
Tris-buffered Saline 0.1% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Immunoprecipitation, Western Blot
Unconjugated
EC 6.3.5.2, Glutamine amidotransferase, GMP synthase, GMP synthase [glutamine-hydrolyzing], GMP synthetase, guanine monphosphate synthetase, guanosine 5'-monophosphate synthase, MLL/GMPS fusion protein
between 650 and 693
20 μg
metabolism
8833
Store at 4°C. Do not freeze.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers