missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Whole Serum Polyclonal Antibody, MP Biomedicals
Brand: MP Biomedicals 0855136
This item is not returnable.
View return policy
Description
Host Species: Goat
Species Reactivity: Human
Specifications
Whole Serum | |
Unconjugated | |
2 mL | |
Human |
Polyclonal | |
Goat | |
Primary | |
Lyophilized |