missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 430, Invitrogen™
Goat Polyclonal Secondary Antibody
Brand: Invitrogen A11063
228.67 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Product will be shipped at Room Temperature.
Mouse IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Mouse IgG (H+L) Cross-Adsorbed in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry.Specifications
Mouse IgG (H+L) Cross-Adsorbed | |
Polyclonal | |
Alexa Fluor 430 | |
Goat | |
purified | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
2 mg/mL | |
PBS with 5mM sodium azide; pH 7.5 | |
Gamma Immunoglobins Heavy and Light chains | |
500 μL | |
Secondary | |
4° C, store in dark | |
IgG |