missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, APC

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen™ A10931

288.01 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.


Product Code. 10707474

  • 432.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Allophycocyanin, Crosslinked, Goat Anti-Rabbit IgG (H+L) is prepared from crosslinked allophycocyanin (APC) and an affinity-purified goat anti-rabbit IgG antibody. Since this phycobiliprotein is optimally excited at 633 nm, it can also be excited at 594 nm; therefore, it can be used with samples simultaneously labeled with Texas Red dye or Alexa Fluor 594 dye without the need of a second excitation source. APC can be used in flow cytometry and imaging applications and is more photostable than Cy5 conjugates.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rabbit IgG (H+L) Cross-Adsorbed
Polyclonal
APC
Goat
purified
RUO
Rabbit
Secondary Antibody
IgG
Flow Cytometry, Immunocytochemistry
1 mg/mL
PBS with 5mM sodium azide; pH 7.5
Gamma Immunoglobins Heavy and Light chains.
500 μL
Secondary
4°C, store in dark
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, APC > 500 μL; APC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.