missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Rabbit IgG (H+L) Secondary Antibody, FITC

Goat Polyclonal Secondary Antibody

Brand:  Invitrogen™ 656111

134.67 EUR valid until 2025-03-29
Use promo code "24111" to get your promotional price.


Product Code. 10682184

  • 202.00 € / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Goat anti-Rabbit IgG (H+L) Secondary Antibody for Western Blot, ICC/IF, IHC (P), IP, ELISA

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rabbit IgG (H+L)
Polyclonal
FITC
Goat
Affinity chromatography
RUO
Rabbit
Secondary Antibody
IgG
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, ELISA, Immunocytochemistry
1.2 mg/mL
PBS with 1% BSA, 40% glycerol and 0.1% sodium azide; pH 7.4
Gamma Immunoglobin.
2 mL
Secondary
4°C, store in dark
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Goat anti-Rabbit IgG (H+L) Secondary Antibody, FITC > 2 mL; FITC

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.