missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgM mu heavy chain,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9406.2mL
This item is not returnable.
View return policy
Specifications
Goat anti-Rabbit IgM mu heavy chain | |
Polyclonal | |
Goat anti-rabbit IgG recognizes rabbit IgM mu heavy chain. This secondary antibody was purified using delipidation and defibrination. | |
Lyophilized from 20mM potassium phosphate, 150mM NaCl, pH 7.2 (0.01% (w/v) sodium azide) | |
Rabbit IgM mu heavy chain | |
2 mL | |
No reaction was observed against rabbit IgG. | |
Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Goat | |
This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against rabbit IgM and rabbit serum. | |
Secondary | |
Rabbit | |
Lyophilized |
For Research Use Only