Learn More
3M™ GoggleGear™ 500 Series Safety Goggles
Scotchgard Anti-Fog coating resists fogging longer than traditional anti-fog coatings and retains its effectiveness even after multiple washings.
Brand:  3M™ 7100074368
Product Code. 15526565
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Can be disinfected with diluted bleach soaking or alcohol wipes without losing its anti-fog performance
  • Offers excellent protection against UV radiation
  • Adjustable elastic strap
  • Indirect ventilation
  • Low profile frame/shroud
  • Polycarbonate lens