Learn More
Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis), Thermo Scientific Chemicals
Form: Wire cloth 100 x 100mm Approx.4.9g; Open area: 56.3%; Width of opening: 0.0075 in.
466.32 € - 915.56 €
Chemical Identifiers
CAS | 7440-57-5 |
---|---|
Molecular Formula | Au |
Molecular Weight (g/mol) | 196.97 |
MDL Number | MFCD00003436 |
InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
PubChem CID | 23985 |
ChEBI | CHEBI:30050 |
IUPAC Name | gold |
SMILES | [Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11365237
|
Thermo Scientific Alfa Aesar
040931.FF |
25 x 25 mm |
466.32 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400351
|
Thermo Scientific Alfa Aesar
040931.FI |
50 x 50 mm |
915.56 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gold gauze, 100 mesh woven from 0.064mm (0.0025in) dia wire, 99.99% (metals basis) is used as an electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Specifications
Golden | |
Gold colored gauze | |
99.99% (Metals basis) | |
MFCD00003436 | |
Insoluble in water. | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Odorless | |
Gold |
7440-57-5 | |
19.32g/cm3 at 20°C | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
100 Mesh | |
Wire Cloth |
Safety and Handling
missing translation for 'einecsNumber' : 231-165-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.