missing translation for 'onlineSavingsMsg'
Learn More

Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis), Thermo Scientific Chemicals

Form: Wire cloth; Open area: 62.7%; Width of opening: 0.015 in.

365.82 € - 2665.26 €

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11345237
SDS View Documents
Thermo Scientific Alfa Aesar
040930.FF
25 x 25 mm
365.82 €
Each
Please to purchase this item. Need a web account? Register with us today!
11490341
SDS View Documents
Thermo Scientific Alfa Aesar
040930.FI
50 x 50 mm
735.66 €
Each
Please to purchase this item. Need a web account? Register with us today!
15434835
SDS View Documents
Thermo Scientific Alfa Aesar
040930.FY
50 x 100 mm
1436.15 €
Each
Please to purchase this item. Need a web account? Register with us today!
11335237
SDS View Documents
Thermo Scientific Alfa Aesar
040930.GH
100 x 100 mm
2665.26 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gold gauze, 52 mesh woven from 0.102mm (0.004in) dia wire, 99.99% (metals basis) is used as an electrode.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

Golden
Gold colored gauze
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
52 Mesh
Wire Cloth
7440-57-5
99.99% (Metals basis)
MFCD00003436
Soluble in aqua regia.
[Au]
196.97
CHEBI:30050
Odorless
Gold
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-165-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.