Learn More
Gold gauze, 82 mesh woven from 0.06mm (0.0025in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Form: Wire cloth; Open area: 63.2%; Width of opening: 0.0097 in.
405.00 € - 804.00 €
Chemical Identifiers
CAS | 7440-57-5 |
---|---|
Molecular Formula | Au |
Molecular Weight (g/mol) | 196.97 |
MDL Number | MFCD00003436 |
InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
PubChem CID | 23985 |
ChEBI | CHEBI:30050 |
IUPAC Name | gold |
SMILES | [Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15484775
|
Thermo Scientific Alfa Aesar
040586.FF |
25 x 25 mm |
405.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348558
|
Thermo Scientific Alfa Aesar
040586.FI |
50 x 50 mm | |||||||
11470311
|
Thermo Scientific Alfa Aesar
040586.FI |
50 x 50 mm |
804.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gold gauze is used to trap mercury vapors during the analysis of mercury by cold vapor-atomic absorption spectrophotometer (CVAAS). It is also used as a surface for condensation of mercury. Further, it is used as an electrode in electrochemistry for the reduction of uranium(V).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Specifications
Golden | |
Gold colored gauze | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
82 Mesh | |
Wire Cloth |
7440-57-5 | |
99.9% (Metals basis) | |
MFCD00003436 | |
Insoluble in water. | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Odorless | |
Gold |
Safety and Handling
missing translation for 'einecsNumber' : 231-165-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only