missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold Germanium foil, 0.1mm (0.004 in.) thick x 43.8mm (1.72in) wide, Premion™, 99.997% (metals basis), Thermo Scientific Chemicals
1540.67 € - 2628.00 €
Specifications
Chemical Name or Material | Gold Germanium foil |
---|---|
Name Note | 0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide |
Avg. Mol. Wt. or Mol. Wt. Range | Au:Ge; 88:12 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD02091706 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313398
|
Thermo Scientific Alfa Aesar
036313.BS |
10 cm |
1540.67 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11323398
|
Thermo Scientific Alfa Aesar
036313.BW |
50 cm | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gold Germanium foil | |
Au:Ge; 88:12 wt% | |
MFCD02091706 | |
Premion™ | |
≈6.45g/10cm |
0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide | |
(metals basis) | |
99.997% | |
Odorless | |
Foil |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only