missing translation for 'onlineSavingsMsg'
Learn More

Gold Germanium slug, 6.35mm (0.25in)dia x 6.35mm (0.25 in.) length, Premion™, 99.999% (metals basis), Thermo Scientific™

1303.00 €

Specifications

Chemical Name or Material Gold Germanium slug
Name Note 6.35mm (0.25 in.)dia. x 6.35mm (0.25 in.) length
Avg. Mol. Wt. or Mol. Wt. Range Au:Ge; 88:12 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD02091706
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15493495
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014173.KJ
5 Pc. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Gold Germanium slug
Au:Ge; 88:12 wt%
MFCD02091706
Premion™
≈2.9g/slug
6.35mm (0.25 in.)dia. x 6.35mm (0.25 in.) length
(metals basis)
99.999%
Odorless
Slug
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications