Learn More
Gold(I) chloride, Premion™, 99.99% (metals basis), Au 84.2% min, Thermo Scientific Chemicals
95.17 € - 1169.82 €
Chemical Identifiers
CAS | 10294-29-8 |
---|---|
Molecular Formula | AuCl |
Molecular Weight (g/mol) | 232.42 |
MDL Number | MFCD00046175 |
InChI Key | FDWREHZXQUYJFJ-UHFFFAOYSA-M |
Synonym | gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special gradeShow More |
PubChem CID | 27366 |
ChEBI | CHEBI:30078 |
IUPAC Name | chlorogold |
SMILES | [Cl-].[Au+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324038
|
Thermo Scientific Alfa Aesar
040432.01 |
0.25 g |
95.17 €
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304038
|
Thermo Scientific Alfa Aesar
040432.03 |
1 g |
264.32 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314038
|
Thermo Scientific Alfa Aesar
040432.06 |
5 g |
1169.82 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
As intermediates. Also In Chemicals, pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-29-8 | |
232.42 | |
FDWREHZXQUYJFJ-UHFFFAOYSA-M | |
27366 | |
chlorogold |
AuCl | |
MFCD00046175 | |
gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special gradeShow More | |
CHEBI:30078 | |
[Cl-].[Au+] |
Specifications
289°C (decomposition) | |
Powder | |
MFCD00046175 | |
14,4515 | |
Very slightly soluble in water. soluble in HCl,HBr organic solvents. | |
[Cl-].[Au+] | |
232.42 | |
CHEBI:30078 | |
99.99% | |
Au 84.2% min. | |
Moisture sensitive | |
(metals basis) |
10294-29-8 | |
AuCl | |
UN3260 | |
gold i chloride, gold monochloride, gold chloride, aurous chloride, gold chloride trihydride, acmc-1bvpa, gold monochloride, acid, gold i chloride trace metals basis, gold i chloride, saj special grade | |
FDWREHZXQUYJFJ-UHFFFAOYSA-M | |
chlorogold | |
27366 | |
232.42 | |
Premion™ | |
Odorless | |
7.57 g/mL | |
Gold(I) chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P234-P260-P264b-P272-P280-P301+P330+P331-P302+P352-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H317-H500
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 233-655-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.