missing translation for 'onlineSavingsMsg'
Learn More

Gold(I) sodium thiosulfate hydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11364707
Click to view available options
Quantity:
1 g
5 g
25 g

Gold(I) sodium thiosulfate hydrate is useful in organogold compounds. Further, it serves as an antirheumatic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15283-45-1
Molecular Formula AuNa3O6S4
Molecular Weight (g/mol) 490.17
MDL Number MFCD00046176
InChI Key FJCMCHNDCIYVST-UHFFFAOYSA-J
Synonym sodium aurothiosulfate, sodium gold thiosulfate, aurothiosulfate, sanochrysin, sanocrysine, crition, crytion, sanocrysin, aurate 3-, bis monothiosulfato-, trisodium, bis monothiosulfato 2--o,s aurate 3-trisodium t-4
PubChem CID 27176
IUPAC Name gold(1+) trisodium disulfanidesulfonate
SMILES [Na+].[Na+].[Na+].[Au+].[O-]S([S-])(=O)=O.[O-]S([S-])(=O)=O

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 1 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Soluble in water. Insoluble in ethanol and most other organic solvents.
Formula Weight 490.19 (anhydrous)
Appearance White powder
Density 3.09
Chemical Name or Material Gold(I) sodium thiosulfate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.