missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold(III) acetate, 99.9% (metals basis), Thermo Scientific Chemicals
146.00 € - 1106.00 €
Chemical Identifiers
CAS | 15804-32-7 |
---|---|
Molecular Formula | C6H9AuO6 |
Molecular Weight (g/mol) | 374.099 |
MDL Number | MFCD00210614 |
InChI Key | OTCKNHQTLOBDDD-UHFFFAOYSA-K |
Synonym | gold 3+ triacetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1 |
PubChem CID | 15952561 |
IUPAC Name | gold(3+);triacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11314717
|
Thermo Scientific Alfa Aesar
039742.02 |
0.5 g |
146.00 €
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11394707
|
Thermo Scientific Alfa Aesar
039742.03 |
1 g |
244.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304717
|
Thermo Scientific Alfa Aesar
039742.06 |
5 g |
1106.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in solar energy technologies. Used in petrochemical cracking and automotive catalysts, water treatment, plating, textiles, research and in optic, laser, crystal and glass applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15804-32-7 | |
374.099 | |
OTCKNHQTLOBDDD-UHFFFAOYSA-K | |
15952561 | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3] |
C6H9AuO6 | |
MFCD00210614 | |
gold 3+ triacetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1 | |
gold(3+);triacetate |
Specifications
15804-32-7 | |
Powder | |
C6H9AuO6 | |
gold 3+ triacetate, gold iii acetate, gold acetate, acmc-1c3i2, triacetic acid gold iii salt, acetic acid, gold 3+ salt 3:1 | |
OTCKNHQTLOBDDD-UHFFFAOYSA-K | |
gold(3+);triacetate | |
15952561 | |
Brown powder |
Brown | |
99.9% (Metal basis) | |
MFCD00210614 | |
Slightly soluble in water. | |
CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Au+3] | |
374.099 | |
374.1 | |
Gold(III) acetate |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only