missing translation for 'onlineSavingsMsg'
Learn More

Gold(III) bromide, 99%, Thermo Scientific Chemicals

140.70 € - 648.23 €

Chemical Identifiers

CAS 10294-28-7
Molecular Formula AuBr3H3
Molecular Weight (g/mol) 439.70
MDL Number MFCD00014171
InChI Key KJCSJLXVPHOFJC-UHFFFAOYSA-N
IUPAC Name gold(III) bromide
SMILES Br.Br.Br.[Au]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11404087
View Documents
Thermo Scientific Alfa Aesar
A19514.03
1 g
140.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
11414087
View Documents
Thermo Scientific Alfa Aesar
A19514.06
5 g
648.23 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Specifications

Chemical Identifiers

10294-28-7
439.70
KJCSJLXVPHOFJC-UHFFFAOYSA-N
Br.Br.Br.[Au]
AuBr3H3
MFCD00014171
gold(III) bromide

Specifications

∼160°C (decomposition)
AuBr3H3
UN3260
KJCSJLXVPHOFJC-UHFFFAOYSA-N
gold(III) bromide
436.69
Hygroscopic
10294-28-7
MFCD00014171
Slightly soluble in water.
Br.Br.Br.[Au]
439.70
99%
Gold(III) bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 233-654-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only