Learn More
Gold(III) bromide, Premion™, 99.99% (metals basis), Au 44.6% min, Thermo Scientific Chemicals
137.00 € - 392.30 €
Chemical Identifiers
CAS | 10294-28-7 |
---|---|
Molecular Formula | AuBr3H3 |
Molecular Weight (g/mol) | 439.70 |
MDL Number | MFCD00014171 |
InChI Key | KJCSJLXVPHOFJC-UHFFFAOYSA-N |
IUPAC Name | gold(III) bromide |
SMILES | Br.Br.Br.[Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11326538
|
Thermo Scientific Alfa Aesar
012162.03 |
1 g |
137.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336538
|
Thermo Scientific Alfa Aesar
012162.06 |
5 g | |||||||
Description
Gold(III) bromide is used as a catalyst in a variety of reactions. It catalyzes the reaction between an enynal unit and carbonyl compounds to form a six-membered cyclic compound. Another catalytic use of gold tribromide is in the nucleophilic substitution reaction of propargylic alcohols, with the gold complex acting as an alcohol-activating agent to facilitate the substitution. It can be used as a testing reagent for the presence of ketamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10294-28-7 | |
439.70 | |
KJCSJLXVPHOFJC-UHFFFAOYSA-N | |
Br.Br.Br.[Au] |
AuBr3H3 | |
MFCD00014171 | |
gold(III) bromide |
Specifications
∼160°C (decomposition) | |
Granules | |
MFCD00014171 | |
Slightly soluble in water. | |
Br.Br.Br.[Au] | |
439.70 | |
99.99% | |
Au 44.6% min. | |
Gold(III) bromide |
10294-28-7 | |
AuBr3H3 | |
UN3260 | |
KJCSJLXVPHOFJC-UHFFFAOYSA-N | |
gold(III) bromide | |
436.69 | |
Premion™ | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 233-654-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only