missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold(III) hydroxide, Au 79% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
10 g
Description
Gold(III) hydroxide is used in medicine, porcelain making, gold plating, and daguerrotypes. Used for preparation of gold catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Powder |
Quantity | 1 g |
Merck Index | 14,4523 |
Solubility Information | Insoluble in water. |
IUPAC Name | gold;trihydrate |
Formula Weight | 247.99 |
Concentration or Composition (by Analyte or Components) | Au 79% min. |
Odor | Odorless |
Sensitivity | Light sensitive |
Chemical Name or Material | Gold(III) hydroxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'