Learn More
Gold(III) hydroxide, Au 79% min, Thermo Scientific Chemicals
Gold plating solutions, porcelain decoration
217.00 € - 2003.00 €
Chemical Identifiers
CAS | 1303-52-2 |
---|---|
Molecular Formula | AuH6O3 |
Molecular Weight (g/mol) | 251.01 |
MDL Number | MFCD00046173 |
InChI Key | PPOYUERUQZXZBE-UHFFFAOYSA-N |
Synonym | gold iii hydroxide, gold trihydroxide, gold hydroxide au oh 3, auh3o3, gold hydroxide, au oh 3, gold iii hydroxide, au |
PubChem CID | 102103 |
SMILES | O.O.O.[Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11376528
|
Thermo Scientific Alfa Aesar
012159.03 |
1 g |
217.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386528
|
Thermo Scientific Alfa Aesar
012159.06 |
5 g |
1059.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366528
|
Thermo Scientific Alfa Aesar
012159.09 |
10 g |
2003.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gold(III) hydroxide is used in medicine, porcelain making, gold plating, and daguerrotypes. Used for preparation of gold catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1303-52-2 | |
251.01 | |
PPOYUERUQZXZBE-UHFFFAOYSA-N | |
102103 |
AuH6O3 | |
MFCD00046173 | |
gold iii hydroxide, gold trihydroxide, gold hydroxide au oh 3, auh3o3, gold hydroxide, au oh 3, gold iii hydroxide, au | |
O.O.O.[Au] |
Specifications
1303-52-2 | |
AuH6O3 | |
14,4523 | |
Insoluble in water. | |
O.O.O.[Au] | |
251.01 | |
247.99 | |
Odorless | |
Gold(III) hydroxide |
Powder | |
MFCD00046173 | |
gold iii hydroxide, gold trihydroxide, gold hydroxide au oh 3, auh3o3, gold hydroxide, au oh 3, gold iii hydroxide, au | |
PPOYUERUQZXZBE-UHFFFAOYSA-N | |
gold;trihydrate | |
102103 | |
Au 79% min. | |
Light sensitive |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 215-120-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only