Learn More
Thermo Scientific™ GC Injection Port Base Seals for Thermo Scientific™ and Agilent Instruments
Ensure optimal GC analysis by using the exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals.
Brand:  Thermo Scientific™ 290GA083
 View more versions of this product
Product Code. 10219174
  • 80.95 € / Pack of 2
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ensure optimal gas chromatography (GC) analysis by using exceptional sealing and inert properties of Thermo Scientific™ GC Injection Port Base Seals. Precision machined from high grade stainless steel, choose from gold-plated or Siltek-treated base seals. The chemically inert coated surface makes them suitable for analysis of active compounds such as pesticides, polychlorinated biphenyls (PCBs) and phenols. Use these GC Injection Port Base Seals with split/splitless injection ports of thermo Scientific™ TRACE™ 1300 Series GC and Agilent GC 5890/6890/7890 systems.
  • Precision machined to provide exceptional sealing properties
  • Reduces activity (especially useful for analysis of pesticide, PCBs, phenols, etc.)
  • High grade stainless steel provides reproducible seal
  • Direct equivalent to Agilent original part
  • Compatible with all Agilent GC 5890/6890/7890 split/splitless injection ports
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gold Inlet Base Seal
Gold Inlet Base Seal
Single Column Installation (Cross Version)
2 Pack
0.8 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.