missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 100nm, carboxy-functionalized, 3kDa PEGylated, OD50, 572nm absorption, Thermo Scientific Chemicals

246.00 € - 478.38 €

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15474535
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J67153.&I
0.5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15495825
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J67153.&I
0.5 mL
272.36 €
0.50mL
Please to purchase this item. Need a web account? Register with us today!
15484535
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J67153.XF
1 mL
478.38 €
1mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

Pink to Red
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.97
CHEBI:30050
100nm
Liquid
7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Odorless
Light sensitive
Gold nanoparticles, carboxy-functionalized, 3kDa PEGylated, OD: 50, 572nm absorption
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only