Learn More
Gold nanoparticles, 15nm, supplied in 0.1mg/mL sodium citrate with stabilizer, OD1, 520nm absorption, Thermo Scientific Chemicals
Description
For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
- Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
Specifications
Specifications
Color | Pink to Red |
Quantity | 25 mL |
Particle Size | 15nm |
Sensitivity | Light sensitive |
Form | Liquid |
Chemical Name or Material | Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 520nm absorption |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.