missing translation for 'onlineSavingsMsg'
Learn More
Gold nanoparticles, 20nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 524nm absorption, Thermo Scientific™ Product Code.: 15424505

Gold nanoparticles, 20nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 524nm absorption, Thermo Scientific™

Product Code. 15424505
1 mL
Click to view available options
Quantity:
0.5 mL
1 mL
This item is not returnable. View return policy

Product Code. 15424505

Brand: Thermo Scientific Alfa Aesar J67061.XF

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Red
Quantity 1 mL
Particle Size 20nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 524nm absorption
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gold nanoparticles, 20nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 524nm absorption, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.