Learn More
Gold nanoparticles, 30nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 526nm absorption, Thermo Scientific Chemicals
84.52 € - 255.27 €
Chemical Identifiers
CAS | 7440-57-5 |
---|---|
Molecular Formula | Au |
Molecular Weight (g/mol) | 196.97 |
MDL Number | MFCD00003436 |
InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
PubChem CID | 23985 |
ChEBI | CHEBI:30050 |
IUPAC Name | gold |
SMILES | [Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15404575
|
Thermo Scientific Alfa Aesar
J67284.AC |
25 mL |
84.52 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15414575
|
Thermo Scientific Alfa Aesar
J67284.AE |
100 mL |
255.27 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
- Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Specifications
Red | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Light sensitive | |
Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 526nm absorption |
7440-57-5 | |
MFCD00003436 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
30nm | |
Liquid |
Safety and Handling
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only