Learn More
Gold nanoparticles, 70nm, biotin-functionalized, PEGylated, OD50, supplied in purified water, 548 nm absorption, Thermo Scientific™
201.50 € - 568.00 €
Chemical Identifiers
CAS | 7440-57-5 |
---|---|
Molecular Formula | Au |
Molecular Weight (g/mol) | 196.97 |
MDL Number | MFCD00003436 |
InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
PubChem CID | 23985 |
ChEBI | CHEBI:30050 |
IUPAC Name | gold |
SMILES | [Au] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3Show More | |
CHEBI:30050 | |
[Au] |
Specifications
Pink to Red | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Light sensitive | |
Gold nanoparticles, biotin-functionalized, PEGylated, OD: 50, supplied in purified water, 548nm absorption |
7440-57-5 | |
MFCD00003436 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
70nm | |
Liquid |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
Your input is important to us. Please complete this form to provide feedback related to the content on this product.