missing translation for 'onlineSavingsMsg'
Learn More

Gold nanoparticles, 80nm, supplied in 0.1 mg/ml sodium citrate with stabilizer, OD1, 553nm absorption, Thermo Scientific Chemicals

Product Code. 15484525
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Suitable for adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Pink to Red
Quantity 100 mL
Particle Size 80nm
Sensitivity Light sensitive
Form Liquid
Chemical Name or Material Gold nanoparticles, supplied in 0.1mg/mL sodium citrate with stabilizer, OD: 1, 553nm absorption

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.