missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold nanoparticles, 90nm, supplied in 0.1mM PBS 95%, OD1, 564nm absorption, Thermo Scientific Chemicals
84.10 € - 239.00 €
Chemical Identifiers
CAS | 7440-57-5 |
---|---|
Molecular Formula | Au |
Molecular Weight (g/mol) | 196.97 |
MDL Number | MFCD00003436 |
InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
PubChem CID | 23985 |
ChEBI | CHEBI:30050 |
IUPAC Name | gold |
SMILES | [Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15420747
|
Thermo Scientific Alfa Aesar
J67426.AC |
25 mL |
84.10 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15474625
|
Thermo Scientific Alfa Aesar
J67426.AE |
100 mL | |||||||
Description
For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Specifications
Pink to Red | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
[Au] | |
196.97 | |
CHEBI:30050 | |
90nm | |
Liquid |
7440-57-5 | |
MFCD00003436 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
95% | |
Light sensitive | |
Gold nanoparticles, supplied in 0.1mM PBS, OD: 1, 564nm absorption |
Safety and Handling
missing translation for 'tsca' : Yes
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only