missing translation for 'onlineSavingsMsg'
Learn More

Gold Nickel wire, 0.5mm (0.02in) dia, annealed, 99.85% (metals basis), Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 036451.BW

 View more versions of this product

Product Code. 11307918

  • 608.03 € / 50cm

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gold Nickel wire
Au:Ni; 82:18 wt%
MFCD02091707
99.85%
Wire
0.5mm (0.02 in.) dia., Annealed, Temper: soft
(metals basis)
50 cm
≈0.32g/10cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only