Learn More
Gold slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals
Form: approx. 3.35-3.60g/slug, approx. weight
1018.07 € - 4182.81 €
Chemical Identifiers
CAS | 7440-57-5 |
---|---|
Molecular Formula | Au |
Molecular Weight (g/mol) | 196.97 |
MDL Number | MFCD00003436 |
InChI Key | PCHJSUWPFVWCPO-UHFFFAOYSA-N |
Synonym | colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 |
PubChem CID | 23985 |
ChEBI | CHEBI:30050 |
IUPAC Name | gold |
SMILES | [Au] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15454755
|
Thermo Scientific Alfa Aesar
014174.KG |
1 Pc. |
1018.07 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15464755
|
Thermo Scientific Alfa Aesar
014174.KJ |
5 Pc. |
4182.81 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gold slugs are used in thermal evaporation and electrodes (quartz oscillator, light-emitting diodes, and laser diodes).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-57-5 | |
196.97 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
23985 | |
gold |
Au | |
MFCD00003436 | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
CHEBI:30050 | |
[Au] |
Specifications
Golden | |
Gold colored slug | |
99.999% (Metals basis) | |
MFCD00003436 | |
Soluble in hot sulfuric acid and aqua regia. Insoluble in water and acid. | |
[Au] | |
196.97 | |
CHEBI:30050 | |
Slug |
7440-57-5 | |
19.32g/cm3 at 20°C | |
Au | |
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3 | |
PCHJSUWPFVWCPO-UHFFFAOYSA-N | |
gold | |
23985 | |
Odorless | |
Gold |
Safety and Handling
missing translation for 'einecsNumber' : 231-165-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.