missing translation for 'onlineSavingsMsg'
Learn More

Gold splatter, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11376627
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]

Specifications

Color Golden
Appearance Gold colored splatters
Assay Percent Range 99.9% (Metals basis)
Quantity 1 g
Odor Odorless
Form Splatters
Chemical Name or Material Gold

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'