missing translation for 'onlineSavingsMsg'
Learn More

Gold standard solution, for AAS, 1mg/mL Au in 0.5N HCl, Thermo Scientific Chemicals

49.85 €

Specifications

Chemical Name or Material Gold standard solution
Concentration 990 to 1010μg/mL of Au(3+) in 10 to 20% HCl
Name Note 1 mg/ml Au in 0.5N HCl
CAS 7440-57-5, 7647-01-0, 7732-18-5
Packaging Plastic bottle
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10666212
View Documents
Thermo Scientific Acros
196001000
100 mL Plastic bottle
49.85 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
Specifications

Specifications

Gold standard solution
1 mg/ml Au in 0.5N HCl
Plastic bottle
Au
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
196.97
1.0900g/mL
Soluble in water
990 to 1010μg/mL of Au(3+) in 10 to 20% HCl
7440-57-5, 7647-01-0, 7732-18-5
Orange/Yellow to Yellow
MFCD00003436
[Au]
196.97
CHEBI:30050
AAS
1.09
Powder and/or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals. Causes severe skin burns and eye damage. May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Danger

Product Identifier
  • Gold standard solution, 1 mg/ml Au in 10-20% HCl

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.