missing translation for 'onlineSavingsMsg'
Learn More

Gold Tin eutectic foil, 0.1mm (0.004 in.) thick x 43.8mm (1.72in) wide, 99.99% (metals basis), Thermo Scientific Chemicals

1394.94 €

Specifications

Chemical Name or Material Gold Tin eutectic foil
Name Note 0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide
Avg. Mol. Wt. or Mol. Wt. Range Au:Sn; 80:20 wt%
Assay Percent Notes (metals basis)
MDL Number MFCD02091709
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383388
View Documents
Thermo Scientific Alfa Aesar
036310.BS
10 cm
1394.94 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Gold Tin eutectic foil is used in electronic manufacturing. It can be made into dimensions much smaller than soft solders, thereby it is utilized in applications which require the highest level of precision.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifica

Specifica

Gold Tin eutectic foil
Au:Sn; 80:20 wt%
MFCD02091709
99.99%
Foil
0.1mm (0.004 in.) thick x 43.8mm (1.72 in.) wide
(metals basis)
Insoluble in water.
≈6.37g/10cm
Videos
Sicurezza e movimentazione

Sicurezza e movimentazione

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documenti

Documenti

Certificati

RUO – Research Use Only