missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 0.35mm (0.0138in) dia, Premion™, 99.999% (metals basis), Thermo Scientific Chemicals

Form: Approx.1.82g/m

186.00 € - 2251.00 €

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15466725
View Documents Promotion Details
Thermo Scientific Alfa Aesar
045084.BU
25 cm
186.00 €
25cm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15476725
View Documents Promotion Details
Thermo Scientific Alfa Aesar
045084.BY
100 cm
669.00 €
100cm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15412147
View Documents Promotion Details
Thermo Scientific Alfa Aesar
045084.CC
500 cm
2251.00 €
500cm
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

Golden
Gold colored wire
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.97
CHEBI:30050
Wire
7440-57-5
99.999% (Metals basis)
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
Odorless
Gold
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-165-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only