missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold wire, 14kt, yellow, 1.63mm (0.064 in.) dia., Au 58.3% min, Thermo Scientific Chemicals
476.37 € - 954.00 €
Specifications
Chemical Name or Material | Gold wire (14kt) |
---|---|
Color | Yellow |
Name Note | 1.63mm (0.064 in.) dia. |
MDL Number | MFCD00003436 |
Solubility Information | Soluble in aqua regia,nitric acid and hydrochloric acid. Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11333898
|
Thermo Scientific Alfa Aesar
040329.BS |
10 cm |
476.37 €
10cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11353898
|
Thermo Scientific Alfa Aesar
040329.BU |
25 cm |
943.70 €
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343898
|
Thermo Scientific Alfa Aesar
040329.BW |
50 cm | N/A | ||||||
Description
Gold wire is used in jewelry and as a plated coating on a wide variety of electrical and mechanical components. It is also acts as a heat reflecting coating for glass as well as a decorative medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gold wire (14kt) | |
1.63mm (0.064 in.) dia. | |
Soluble in aqua regia,nitric acid and hydrochloric acid. Insoluble in water. | |
≈28g/m |
Yellow | |
MFCD00003436 | |
% Concentration Cu>Zn>Ag, Au ≤58.3% | |
Wire |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only