missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 2.0mm (0.08in) dia, 99.95% (metals basis), Thermo Scientific Chemicals

174.87 € - 3660.21 €

Chemical Identifiers

CAS 7440-57-5
Molecular Formula Au
Molecular Weight (g/mol) 196.97
MDL Number MFCD00003436
InChI Key PCHJSUWPFVWCPO-UHFFFAOYSA-N
Synonym colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
PubChem CID 23985
ChEBI CHEBI:30050
IUPAC Name gold
SMILES [Au]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307058
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014729.BO
1 cm
174.87 €
1cm
Please to purchase this item. Need a web account? Register with us today!
11397048
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014729.BQ
5 cm
872.34 €
5cm
Please to purchase this item. Need a web account? Register with us today!
11387048
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014729.BU
25 cm
3660.21 €
25cm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-57-5
196.97
PCHJSUWPFVWCPO-UHFFFAOYSA-N
23985
gold
Au
MFCD00003436
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
CHEBI:30050
[Au]

Specifications

7440-57-5
MFCD00003436
PCHJSUWPFVWCPO-UHFFFAOYSA-N
gold
23985
99.95%
≈60.6g/m
(metals basis)
Au
colloidal, powder, flake, leaf, gold, colloidal, burnish, shell, ci pigment metal 3, magnesium purple, c.i. pigment metal 3
[Au]
196.97
CHEBI:30050
Odorless
Wire
Gold wire, 2.0mm (0.08 in.) dia.
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-165-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only