missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gold Zinc slugs, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.99% (metals basis), Thermo Scientific™
501.50 € - 1830.00 €
Specifications
Chemical Name or Material | Gold Zinc slugs |
---|---|
Name Note | 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length |
Avg. Mol. Wt. or Mol. Wt. Range | Au:Zn; 95:5 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD02092916 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Gold Zinc slugs | |
Au:Zn; 95:5 wt% | |
MFCD02092916 | |
≈3.7g/slug |
6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length | |
(metals basis) | |
99.99% | |
Slug |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title