missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Autoclave Bags Printed with Biohazard Logo

Product Code. 15567885
Click to view available options
Capacity (Metric):
3 L
9 L
14 L
21 L
33 L
Width (Metric):
255 mm
300 mm
400 mm
600 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15567885

Brand: Corning™ SA3310

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Used for contaminated waste disposal in autoclave. Gosselin™ Autoclave Bags Printed with Biohazard Logo can withstand autoclaving temperatures up to 134°C/273.2°F.

  • Polypropylene (PP) resistant bags
  • Thermal resistance 134°C
  • Transparent
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 33 L
Material Polypropylene
Thickness (Metric) 40 μm
No. per Case 500/Cs.
No. per Pack 1/Pk.
Width (Metric) 600 mm
Height (Metric) 780 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.