missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Autoclave Bags Printed with Biohazard Logo

Used for contaminated waste disposal in autoclave. Gosselin™ Autoclave Bags Printed with Biohazard Logo can withstand autoclaving temperatures up to 134°C/273.2°F.

23.01 € - 105.00 €

Specifications

Material Polypropylene
Thickness (Metric) 40 μm
No. per Case 500/Cs.
No. per Pack 1/Pk.
View More Specs

Products 5
Product Code Brand Capacity (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Width (Metric) Price Quantity & Availability  
15527885
View Documents
Corning™
SA03-10
3 L 255 mm
23.01 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
15537885
View Documents
Corning™
SA09-10
9 L 300 mm
40.85 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
15547885
View Documents
Corning™
SA14-10
14 L 400 mm
63.70 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
15557885
View Documents
Corning™
SA21-10
21 L 400 mm
70.20 €
Pack of 100
Estimated Shipment: 25-02-2025
to see stock.
Add to basket
 
15567885
View Documents
Corning™
SA33-10
33 L 600 mm
105.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Polypropylene (PP) resistant bags
  • Thermal resistance 134°C
  • Transparent
Specifications

Specifications

Polypropylene
40 μm
500/Cs.
1/Pk.
Videos
SDS
Documents

Documents

Product Certifications