missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gosselin™ Flat-Base Tubes, PS
Excellent multi-purpose storage tubes with a cap closure. Gosselin™ Flat-Base Tubes, PS have a flat base to facilitate self standing.
Brand: Gosselin™ FLY35-02
60.52 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polysytrene construction
- Translucent
- Flat bottom for self standing
- Cap closure
Specifications
Flat | |
Natural | |
95 mm | |
Tube |
35 mL | |
Clinical, dairy, environment, industries, microbiology | |
Polystyrene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction