missing translation for 'onlineSavingsMsg'
Learn More

Gosselin™ Serological Graduated Pipettes

Product Code. 15344734
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
25 mL
Quantity:
1000 Pack
1500 Pack
2000 Pack
400 Pack
4000 Pack
800 Pack
Unit Size:
Pack of 1000
Pack of 1500
Pack of 2000
Pack of 400
Pack of 4000
Pack of 800
This item is not returnable. View return policy

Product Code. 15344734

Brand: Gosselin™ GPL10E25

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provides accurate transfer of small amounts of liquid in the laboratory. Gosselin™ Serological Graduated Pipettes are manufactured from clear polystyrene, allowing for ease of readability.

  • Single piece construction
  • Color-coded hydrophobic plug
  • Nonpyrogenic, noncytotoxic and nonhaemolytic
TRUSTED_SUSTAINABILITY

Specifications

Material Polystyrene
For Use With (Application) For dairy, environment, industries, microbiology
Graduated Yes
Quantity 1000 Pack
Volume (Metric) 10 mL
Length (Metric) 294 mm
Type Graduated Pipet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.