missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Sterile Polyethylene Blender Bags
Universal bags for homogeneous suspension
337.35 € - 949.95 €
Product Code | Brand | Capacity (Metric) | Description | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Description | Quantity | Price | Quantity & Availability | |||
11787919
|
Corning™
BBAG-04 |
400 mL | With lateral filter | 50/Pk. |
949.95 €
Pack of 1500 |
In Stock
Log in to see stock. |
|||
11777919
|
Corning™
FBAG-04 |
400 mL | With full filter | 50/Pk. |
922.01 €
Pack of 1500 |
Estimated Shipment: 23-05-2025
Log in to see stock. |
|||
11755085
|
Corning™
SM2-01 |
400 mL | Without filter | 50/Pk. |
337.35 €
Pack of 2500 |
In Stock
Log in to see stock. |
|||
Description
- Polyethylene (PE) bags
- 80 to 3500 mL
- With or without filter
- Traceability on each filter bag
- Sterile
Order Info
Use bag clip, Cat. No. 07-201-979, to close bags between sample preparation steps.
Specifications
Natural | |
Polyethylene | |
Sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title